ContactUsLinkComponent
Yeast Live-or-Dye™ Fixable Live/Dead Staining Kit, Biotium
Yeast Live-or-Dye™ Fixable Live/Dead Staining Kit, Biotium
# de catalogue 75845-626
Fournisseur:  Biotium
Numéro CAS:  
Kit Yeast Fixable Live Dead Staining
Yeast Live-or-Dye™ Fixable Live/Dead Staining Kit, Biotium
# de catalogue 75845-626
Fournisseur:  Biotium
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Color:
    Green/Red
  • Description:
    Yeast live-or-dye fixable live/dead staining kit
  • Size:
    1000 assays
  • Cat. no.:
    75845-626
  • Supplier No.:
    31064
Spécifications
A propos de cet article
The Yeast Live-or-Dye™ Fixable Live/Dead Staining Kit is a fixable, fluorescent assay to determine yeast viability. The kit contains two fluorescent components, Thiazole Orange and Live-or-Dye™ 568/583.
  • Stable and selective dead cell staining
  • Simple, rapid, no-wash staining protocol
  • Tolerates formaldehyde fixation
  • Suitable for flow cytometry and microscopy
Live-or-Dye™ 568/583 is a cell membrane impermeable amine-reactive dye. It is able to enter into dead cells that have compromised membrane integrity and covalently label free amines on intracellular proteins. This dye labeling is extremely stable, allowing the cells to be fixed and permeabilized without loss of fluorescence or dye transfer between cells. It is suitable for detection by fluorescence microscopy or flow cytometry.
Thiazole Orange is a green fluorescent dye that localizes to the cytoplasm, with some nuclear accumulation. It is fixable, resulting in diffuse cytoplasmic staining in fixed cells.
After staining yeast with this kit, both living and dead cells will be stained with green fluorescence. Only dead cells will be stained with red fluorescence.
Spectral Properties
Live-or-Dye™ 568/583 Ex/Em 568/583 nm
Thiazole Orange Ex/Em 512/533 nm
Delivery: Kit includes Live-or-Dye™ 568/583 (solid), four vials, Thiazole Orange 10 mM in DMSO, 100 µl and Anhydrous DMSO, 250 µl
Customers Who Bought This Also Bought