ContactUsLinkComponent
Yeast Vitality Staining Kit, Biotium
Yeast Vitality Staining Kit, Biotium
# de catalogue 75845-618
Fournisseur:  Biotium
Numéro CAS:  
Kit Yeast Vitality Staining 1000Assays
Yeast Vitality Staining Kit, Biotium
# de catalogue 75845-618
Fournisseur:  Biotium
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Color:
    Red/Green/Blue
  • Description:
    Yeast vitality staining kit
  • Size:
    1000 assays
  • Cat. no.:
    75845-618
  • Supplier No.:
    31062
Spécifications
A propos de cet article
A fluorescent vitality dye for yeast, identical in structure to the dye FUN® 1. Living yeast cells show red fluorescent vacuolar tubules.
  • For live-cell imaging of yeast and fungi
  • ViaVac™ Red/Green stains the vacuole red in metabolically active cells
  • Calcofluor White labels all cell walls blue
The Yeast Vitality Staining Kit is a fluorogenic assay that allows you to determine whether your yeast cells are metabolically active. The kit contains two fluorescent components: Calcofluor White (75832-838) and ViaVac™ Red/Green (75832-630).
ViaVac™ Red/Green (75832-630) is a fluorescent viability dye for yeast, identical in structure to the dye FUN® 1. ViaVac™ passively diffuses into cells, showing green cytoplasmic staining. In many species of yeast and fungi, in metabolically active cells the dye is actively transported into the vacuole, resulting in red fluorescent staining of tubular intravacuolar structures.
Calcofluor White (75832-838) is a fluorescent blue dye that binds to cellulose and chitin, which can be found in the cell walls of fungi, algae, and plants.
After staining yeast with this kit, both living and dead cells will be outlined in blue, while only living cells will show red fluorescent, vacuolar tubules.
Spectral Properties
ViacVac™ Red/Green Ex/Em ~485/620 nm (red); 485/530 nm (green)
Calcofluor White Ex/Em 360/430 nm
FUN®1 is a registered trademark of Thermo Fisher Scientific.
Delivery: Kit contains ViaVac™ Red/Green, 10 mM in DMSO, 100 µl and Calcofluor White, 5 mM in water, 500 µl.
Customers Who Bought This Also Bought