Finntip™ Wide Orifice Pipette Tips, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Eliminate cell fragmentation caused by mechanical shearing with Thermo Scientific™ Finntip™ Wide Orifice Pipette Tips.
- Two different models cover volume range of 10 - 1000 μL
- Wide orifice tip; inside diameter of orifice is 1.1 mm/1.8 mm
- Color code: White
- Length: 5.3 cm/7.2 cm
Ideal for pipetting macromolecules.
Customers Who Bought This Also Bought