Gel Packs, Cryopak Verification Technologies
Fournisseur: Cryopak Verification Technologies
Certificats
A propos de cet article
Available in multiple sizes, gel packs are a convenient way to keep temperature sensitive shipments within the desired temperature range.
- Non-to×ic and non-hazardous
- Packaged in durable plastic film
- Freezing/melting point of 0 °C
Cryopak gel packs are a cost-effective way to safely transport items within the cold chain. Ideal for the pharmaceutical and food industries, these packs allow products to maintain their required storage temperatures during transit.
Customers Who Bought This Also Bought