Ecopak Sustainable Shippers, Cryopak Verification Technologies
Supplier: Cryopak Verification Technologies
Certificates
About this item
These sustainable shippers are compostable and recyclable.
- Multiple thickness options
- Protective cushioning
- Replaces EPS products and other insulators
The Cryopak Ecopak liner solutions are manufactured in a zero waste environment from a recycled fiber blend and wrapped in a compostable film that absorbs moisture.
This material is manufactured to ensure that each liner provides consistent thermal performance for all specific shipping requirements.
Customers Who Bought This Also Bought