CryoCube™ Reusable Shippers, Cryopak Verification Technologies
Fournisseur: Cryopak Verification Technologies
Certificats
A propos de cet article
These cost-effective shippers reduce shipping costs by reducing the total weight of shipments.
- Maintains temperature up to 120 hours
- Highly reusable
- Available for 2 to 8 °C and CRT configurations
Vacuum insulated panels provide the highest thermal insulation quality available. The insulated panels are protected with plastic wrap and foam padding for increased durability.
ith increased duration capabilities and a rugged PCOR plastic corrugated outer shipper for increased reusability, the CryoCube™ is the most robust option, maintaining a controlled internal environment for up to 120 hours.
Customers Who Bought This Also Bought