TimeSaver™ Pre-Qualified Shippers, Cryopak Verification Technologies
Supplier: Cryopak Verification Technologies
Certificates
About this item
Pre-qualified shipping solutions are tested to the highest quality standards to preserve the integrity of transported products.
- Packout instructions to ensure performance
- Abridged qualification report
- Qualification certificate
The TimeSaver™ solutions protect products that must be shipped within a specific temperature range for up to 24, 36, or 48 hours.
Customers Who Bought This Also Bought