PUR-Forma Polyurethane Shippers, Cryopak Verification Technologies
Supplier: Cryopak Verification Technologies
Certificates
About this item
Cryopak’s PUR-Forma line of pre-qualified polyurethane shippers provide outstanding thermal insulation, making them an ideal choice for protecting temperature-sensitive shipments from environmental hazards.
- PUR wall thickness of 1.5 to 3"
- E×ceptional strength and durability
- Durable plastic outer skin for reusability
In addition to the low heat conductivity, PUR is rugged and lightweight, which is ideal for return/reuse logistics.
Customers Who Bought This Also Bought