A propos de cet article
The ChromasterUltra Rs is the result of many years of research, development and experience, leading to the introduction of one of the most advanced, robust and easy to use UHPLC system available on the market today. Hitachi-High Technologies is a world leader in fluid control and optical analysis technology, as well as in high performance electronics. Hitachi has focused on combining the knowledge of these three key disciplines to give the world of chromatography one of the best instruments for today's demands. To arrange a demonstration with one of our experts, please email [email protected].
- Very high pressure rating (20305.28 psi) for high resolution columns
- High pressure gradient with integrated solvent selector (2×2)
- Integrated degasser and washing pump
- Extremely sensitive DAD with capillary flow cell technology
- Very low noise and high resolution detectors
- Adaptable cooled autosampler for maximized flexibility
- Big column oven with optional six-column selector
- Integrated waste tank and solvent organizer
For the ChromasterUltra Rs UHPLC system there are many different chromatography software packes available, e.g. Hitachi Chromaster Ultra System Manager, Waters Empower 3 and Agilent Openlab EZChrom Edition.