ContactUsLinkComponent
Innovating Science® STEM Investigations: Design a Biodegradable Plastic
Innovating Science® STEM Investigations: Design a Biodegradable Plastic
# de catalogue 470329-186
Fournisseur:  Ward's Science
Kit
Innovating Science® STEM Investigations: Design a Biodegradable Plastic
# de catalogue 470329-186
Fournisseur:  Ward's Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    STEM Investigations: Design a Biodegradable Plastic Kit, for 15 Groups/Tests
  • Cat. no.:
    470329-186
Spécifications
A propos de cet article
Learn about biplastics and how their composition affects their physical properties.
  • Students will make samples of different bioplastics and design an experiment to compare the biodegradability of their bioplastic samples to a traditional plastic.
  • Aligned to NGSS Practices
  • Performance expectations: HS-PS2-6, HS-LS2-7, HS-ESS3-4, HS-ETS1-1, HS-ETS1-2, HS-ETS1-3
  • Disciplinary core ideas: PS2.B, LS2.C, LS4.D, ETS1.B, ESS3.C, ETS1.A, ETS1.C
  • Cross cutting concepts include structure and function. Stability and change
  • Engineering practices include obtaining evaluating and communicating information, constructing explanations and designing solutions. Asking questions and defining problem
Hundreds of millions of tons of plastic are produced each year and used for a variety of products, from packaging to automobiles. The majority of these products are disposable items, leading to excessive amounts of plastic being discarded into landfills and the oceans. Due to the major threat this poses to the environment, scientists and engineers are developing bioplastics that break down more quickly and produce less harmful byproducts than traditional plastics. In this activity, students will make samples of different bioplastics to study how their composition affects their physical properties. They will then use what they have learned to create a bioplastic that could be used to replace the PET plastic in disposable water bottles. Additionally, students will design an experiment to compare the biodegradability of their bioplastic samples to a traditional plastic. Kit contains enough materials for 15 groups. Teacher's Manual and Student Study Guide copy masters are included.
Delivery: Kit includes instructional manual and SDS booklet.
Customers Who Bought This Also Bought