LDPE Pasteur Pipettes, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Non-toxic All-in-one pipettes.
- Molded
- Unbreakable
- Translucent
- Pack of 500
These pipettes are all non-toxic and molded from unbreakable translucent lowdensity polyethylene. These can be sealed and refrigerated. They are available in sterile and non-sterile forms. The sterile pipettes are individually wrapped.
Customers Who Bought This Also Bought