Disposable PE Transfer Pipettes, United Scientific Supplies
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
All-in-one pipettes.
- Molded
- Unbreakable
- Translucent
- Pack of 100
All-in-one pipettes are molded from unbreakable translucent polyethylene. These versitle pipettes can be sealed and refrigerated. Perfect for classroom and laboratory use.
Customers Who Bought This Also Bought