A propos de cet article
Quickly heat and dry samples.
- Stainless steel inner chamber resists corrosion
- Rounded inner chamber corners offer easy cleaning
- Samples can be heated up to 482 °F (250 °C)
Mechanical convection ovens feature a PID controller which provides accurate and reliable temperature control up to 482 °F (250 °C). Ideal for drying and sterilization. The ovens require minimal heating time, reaching 212 °F (100 °C) in 20 to 50 minutes. Easy-to-view LCD reads time, current temperature, set temperature, running status, and fan status.
Silicone rubber gasket offers a tight seal, ensuring good heat retention and low energy consumption. The gasket has a long service life and can be easily replaced when needed. Additional oven features include auto-restart after power loss/return, temperature deviation alarm, over-temperature set point alarm, and overcurrent protection. An air vent located at the back of the unit can be adjusted with the knob located in front of the oven to control inner air and vapor circulation.
Spécifications
- Catalog No:
- 76492-758
- 76492-760
- 76492-766
- 76492-762
- 76492-764
- Temperature variation:
- ±1.5% at 100 °C
- ±2% at 100 °C
- ±3% at 100 °C
- ±2.5% at 100 °C
- ±3% at 100 °C
- Controller:
- PID temperature controller
- PID temperature controller
- PID temperature controller
- PID temperature controller
- PID temperature controller
- Shelves supplied (max.):
- 2 (4)
- 2 (5)
- 2 (10)
- 2 (6)
- 2 (7)
- Timer:
- 0 - 99 h, 59 min
- 0 - 99 h, 59 min
- 0 - 99 h, 59 min
- 0 - 99 h, 59 min
- 0 - 99 h, 59 min