A propos de cet article
Low energy consumption provides operating cost savings.
- Stainless steel inner chamber resists corrosion
- Rounded inner chamber corners offer easy cleaning
- Samples can be heated up to 392 °F (200 °C)
Gravity convection ovens feature a PID controller which provides accurate and reliable temperature control up to 392 °F (200 °C). Natural convection heating allows sample temperature to be uniform; suitable for sterilization, powder drying, and high-temperature storage. The ovens require minimal heating time, reaching 212 °F (100 °C) in 15 to 30 minutes. Easy-to-view LCD reads time, current temperature, set temperature, running status, and fan status.
Silicone rubber gasket offers a tight seal, ensuring good heat retention and low energy consumption. The gasket has a long service life and can be easily replaced when needed. Additional oven features include auto-restart after power loss/return, temperature deviation alarm, over-temperature set point alarm, and overcurrent protection.
Spécifications
- Catalog No:
- 76492-742
- 76492-744
- 76492-746
- 76492-748
- Temperature variation:
- ±3% at 100 °C
- ±3% at 100 °C
- ±3.5% at 100 °C
- ±3.5% at 100 °C
- Controller:
- PID temperature controller
- PID temperature controller
- PID temperature controller
- PID temperature controller
- Shelves supplied (max.):
- 2 (6)
- 2 (8)
- 2 (10)
- 2 (11)
- Timer:
- 0 - 99 h, 59 min
- 0 - 99 h, 59 min
- 0 - 99 h, 59 min
- 0 - 99 h, 59 min