Deuterium Lamp, Gilson/Rainin 115, 116, 117, Optimize Technologies
# de catalogue 89522-424
Fournisseur: Optimize
Spécifications
- Description:Deuterium lamp, Gilson/Rainin 115, 116, 117
- Cat. no.:89522-424
- Supplier No.:26-32-03671
Spécifications
A propos de cet article
Optimize Technologies offers a broad selection of lamps for OEM HPLC detectors.
- 1000 Hour lifetime guarantee on all Standard Lamps
- 2000 Hour lifetime guarantee on all DX2 DoubleLife
Optimize offers a broad selection of lamps for OEM HPLC detectors. The high quality standard lamps come with a guaranteed 1,000 hour lifetime. We also offer our DX2 DoubleLife» Lamps. These long-life lamps are made using only top quality components and come with a 2,000 hour lifetime guarantee.
Ordering information: Need a lamp that is not listed on our website? Please contact us.
Customers Who Bought This Also Bought