ContactUsLinkComponent
Hollow Acrylic Cube
Hollow Acrylic Cube
# de catalogue 470328-128
Fournisseur:  Wards
Hollow Acrylic Prism
Hollow Acrylic Cube
# de catalogue 470328-128
Fournisseur:  Wards
Spécifications
  • Size:
    25×25×25 mm
  • Cat. no.:
    470328-128
Spécifications
A propos de cet article
Designed to study and demonstrate Snell's Law of Refraction.
  • Made with high quality acrylic plates
  • Edges and points are fully cemented
  • Includes stopper for leak-free use when filled
When referring to light or other waves passing through a boundary between two different isotropic media, Snell's law can be used to describe the relationship between the angles of incidence and refraction. Fill the hollow cube with a liquid of your choice. These are also useful when studying other principles of light manipulation, light rays, and light angles.
Customers Who Bought This Also Bought