ContactUsLinkComponent
Linearity FLQ Urine/Fluids Chemistry for Beckman AU, AUDIT MircoControls
Linearity FLQ Urine/Fluids Chemistry for Beckman AU, AUDIT MircoControls
# de catalogue 76492-630
Fournisseur:  Audit Microcontrols
Numéro CAS:  
Linearity FLQ Urine/Fluids Chemistry for Beckman Au, Format: Frozen Liquid, Storage: 8 hours when stored at 2-8 deg C, Size: 3ml
Linearity FLQ Urine/Fluids Chemistry for Beckman AU, AUDIT MircoControls
# de catalogue 76492-630
Fournisseur:  Audit Microcontrols
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Linearity FLQ urine/fluids chemistry for Beckman AU
  • Cat. no.:
    76492-630
  • Volume:
    3 ml
  • Includes:
    10 vials
Spécifications
A propos de cet article
The Audit® MicroControlsTM Linearity FLQ Urine/Fluids Chemistry for Beckman AU is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: Amylase, Calcium, Chloride, Creatinine, Glucose, Magnesium, Microalbumin, Microprotein, Potassium, Sodium, Phosphorous, Urea Nitrogen, Uric Acid.
Customers Who Bought This Also Bought