Brady® Thermal Transfer 1 Core R6000 Halogen-Free Series Printer Ribbon
Fournisseur: Brady Worldwide
Certificats
A propos de cet article
This ink-side-out ribbon features an open 1" core for compatibility with a wide variety of thermal transfer printers.
- Halogen-free
- Heat-resistant
- Scratch-resistant
- Solvent-resistant
One-color ribbon for printing a single color onto a label. R6000 Series Halogen-Free Resin Formulation offers excellent solvent, scratch and heat resistance when printed on the appropriate label material. Recommended for printing on glossy or very smooth label stocks, such as B-483 Ultra aggressive polyester and B-423 Permanent polyester from Brady®. Combine Brady label materials with their recommended ribbon series for superior legend performance.
These ribbons are designed for use with BBP®81 Label Printer, BradyPrinter i7100 Industrial Label Printer, and THT 3" Core Printers.
Customers Who Bought This Also Bought