Brady® i5100 IP 1 Core R6200 Series Printer Ribbon
Fournisseur: Brady Worldwide
Certificats
A propos de cet article
IP-Enabled ribbons come spooled on a specially designed core that includes a Smart Cell RFID tag that communicates with the Brady IP® industrial label printer.
- Scratch-resistant
- Smear-resistant
- Chemical-resistant
- Heat-resistant
One-color ribbon for printing a single color onto a label. R6200 Series Ribbons offer superior scratch, smear, heat and chemical resistance when printed on the appropriate label material from Brady®. Recommended for printing on glossy, smooth or semi-matte label stock, such as B-498 repositionable vinyl cloth and B-486 metallized polyester. The Brady IP® industrial label printer status monitor shows the printer and material information on the user's PC.
These ribbons are designed for use with Brady IP® Series Label Printer and BradyPrinter i5100 Industrial Label Printer.
Customers Who Bought This Also Bought