Brady® i5100 IP 1 Core R4300 Series Printer Ribbon
Fournisseur: Brady Worldwide
Certificats
A propos de cet article
IP-Enabled ribbons come spooled on a specially designed core that includes a Smart Cell RFID tag that communicates with the Brady IP® industrial label printer.
- Chemical-resistant
- Smear-resistant
One-color ribbon for printing a single color onto a label. R4300 Series Ribbons offers strong environmental resistance against smearing and chemicals when printed on the appropriate Brady® label material. Recommended for printing on paper and films that have a matte or rough surface, such as B-427 Self-laminating vinyl and B-499 Nylon cloth from Brady®. The Brady IP® industrial label printer status monitor shows the printer and material information on the user's PC.
These ribbons are designed for use with Brady IP® Series Label Printer and BradyPrinter i5100 Industrial Label Printer.
Customers Who Bought This Also Bought