Heat Shrink Polyolefin Wire Marking Sleeves, Jumbo Roll, Brady
Fournisseur: Brady Worldwide
Certificats
A propos de cet article
PermaSleeve® Heat Shrink Tubing is engineered for thermal transfer or dot matrix printing and is ideal for use in wire identification.
- Flame-retardant
- Heat-shrinkable
- Wire diameter range: 0.094 to 0.215"
Sleeves are designed for use with BradyPrinter i7100 Industrial Label Printer, BSP45 Wire and Cable Sleeve Applicator, and THT 3" Core Printers.
Ordering information: A ribbon is required to print this material. Use Part Number IP-R6606 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use R6606 for a different printer.
Customers Who Bought This Also Bought