Heatex Polyolefin 20 to 16 Gauge Wire Marking Sleeves, 3" Core, Brady
Fournisseur: Brady Worldwide
Certificats
A propos de cet article
Heat-Shrink PermaSleeve® HX Polyolefin (B-7642) Wire Marking Sleeves are 2:1 heat shrinkable, high-temperature rated, highly flame retardant, and very flexible.
- Heat-shrinkable
Heat-Shrink PermaSleeve® HX Polyolefin (B-7642) Wire Marking Sleeves are supplied in roll form in a flattened format on a carrier designed for use with computer driven printers. Operating temperature of ‒40 to +120 °C (‒40 to +248 °F).
Sleeves are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®72 Label Printer, BradyPrinter i7100 Industrial Label Printer, and THT 3" Core Printers.
Certifications: Meets SAE-AMS-DTL-23053/5 Class 1 & 3, SAE-AS-81531, MIL-STD-2002. Tested per ASTM E162 and ASTM E662.
Ordering information: A ribbon is required to print this material. Use Part Number 490006-292.
Customers Who Bought This Also Bought