Self-Laminating Vinyl Wire and Cable Labels, 3" Core, Brady
Fournisseur: Brady Worldwide
Certificats
A propos de cet article
Self-Laminating Vinyl (B-427) is an excellent material for wire and cable identification. This material has good clarity, conformability, and is self-extinguishing.
- Flame-retardant, self-laminating material
- Service temperature: ‒40 to +70 °C (‒40 to +158 °F)
Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text. Durable, permanent acrylic adhesive has outstanding water and oil resistance.
Labels are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, or THT 3" Core Printers.
Ordering information: A ribbon is required to print this material. Use Part Number 89079-927 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use 49006-270 for a different printer.
Customers Who Bought This Also Bought