Reagent Reservoirs, Globe Scientific
Fournisseur: Globe Scientific
Certificats
A propos de cet article
These reagent reservoirs are suitable for virtually all of your lab’s sample preparation and aliquoting needs.
- Pour spouts at every corner
- Rigid sidewall design
- Wide, stable base
- Molded graduations
- Compatible with multi-channel pipettors
- Dnase, Rnase, purogen, animal materials and latex free
- Produced from virgin medical grade polystyrene
Design enhancements include pour spouts at each corner, a rigid sidewall design and graduations for easy volume reference. Sterile variants are irradiated to guarantee a sterility assurance level (SAL) of 10-⁶. Available volumes of 10 ml, 25 ml and 50 ml ensure there is a volume and packaging option to fit almost every application. Individual, bulk and five-pack options are available.
Customers Who Bought This Also Bought