CoolCube™ Microtube and PCR Plate Cooler
# de catalogue 470334-084
Fournisseur: Ward's
Spécifications
- Capacity:36× 1.5/2.0 ml tubes (side 1); 96× 0.2 ml tubes; 10× 1.5/2.0 ml tubes (side 2)
Spécifications
A propos de cet article
The CoolCube™ stores samples safely on the lab bench preventing degradation from temperature fluctuation.
- Maintains 0 °C for up to 4 hours
- Also for use as a PCR workstation
- Two sides, ideal for: 1.5 and 2.0 ml microtubes, 0.2 ml PCR tubes and strips, 96 well PCR plates
Simply place it in the freezer overnight and it’s ready for use.
Customers Who Bought This Also Bought