Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Fournisseur: Corning
Certificats
A propos de cet article
75 mm Transwell® with 0.4 µm pore polycarbonate membrane insert, sterile.
- 10 µm thick translucent polycarbonate membrane
- Treated for optimal cell attachment
- Each insert is packaged in a 100 mm dish, 12 inserts per case
- Membrane must be stained for cell visibility
Produces a cell culture environment that closely resembles an in vivo state. An essential tool for the study of both anchorage-dependent and independent cell lines.
Spécifications
- Catalog No:
- 76404-722
- 29442-130
- 29442-088
- 29442-086
- 29442-106
- 29442-104
- 29442-110
- 29442-120
- 29442-108
- 29442-118
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk