Justrite® Safety Drum Pumps for 30 and 55 Gallon Drums
Fournisseur: Justrite
Certificats
A propos de cet article
Ideal for small quantity dispensing of liquids.
- For 30 and 55 gal. drums
- 8 oz. per stroke
- 36" reach below lid
- 22" suction tube length
This manually operated piston pump is FM approved and delivers up to 8 oz. per stroke, ideal for transferring flammable and combustible liquids in a controlled and safe manner. PTFE seals, offering superior chemical resistance and a long life. Included with the pump is a standard 2" NPT threaded bung adapter. The included pick-up tube allows the pump to reach up to 36" below the lid of the drum. Included flash arrestor keeps contents safe from external flame flashbacks. The bonding and grounding clamps come pre-assembled and provide the means to safely ground the receiving container to avoid a static discharge.
FM approved.
Ordering information: One-year warranty.
Delivery: Includes pump, pick-up tube, 2" NPT bung adapter, grounding wires with clamps, instruction sheet.
Spécifications
- Catalog No:
- 76474-344
- 76474-348
- Model:
- Model 07712 (Electro zinc plated steel)
- Model 07717 (Stainless steel)
- Connection:
- 2" NPT (connection to container)
- 2" NPT (connection to container)
- Flow rate:
- 8 oz./stroke
- 8 oz./stroke
- Material:
- Electro zinc plated steel flame arrestor, 0.5"
- Stainless steel flame arrestor, 0.5"
- Max. viscosity:
- 90 Wt.
- 90 Wt.
- Outlet:
- 0.5" Ø spout
- 0.5" Ø spout
- Wetted materials:
- Electro zinc plated steel and PTFE seals
- Stainless steel and PTFE seals