VWR® Plus Series Undercounter Laboratory Freezers Freestanding (–20 °C)
# de catalogue 76488-176
Fournisseur: VWR International
Spécifications
- Temperature Range:–15...–25 °C
- Description:Undercounter laboratory freezer, freestanding, solid door (–20 °C)
- Environmentally Preferable:Safer Human and Environmental Health,Energy Efficient
- Cat. No.:76488-176
- Electrical:115 V, 60 Hz, 1.4 Amps, ¹/₈ HP
- Defrost:Manual
- Door type:1 solid swing door, right hinged
- Shelves/drawers:2 fixed shelves
- Weight:58 kg (128 lbs.)
- Exterior W×D×H:600×616×815 mm (23⅝×24¼×32³/₁₆")
- Interior W×D×H:483×406×660 mm (19×16×26")
- Capacity:113 L (4 cu.ft.)
Spécifications
A propos de cet article
The compact undercounter laboratory freezer provides quality cold storage in limited-space settings.
- Microprocessor temperature controller
- Digital temperature display
- Audible and visual high and low temperature alarms
- Remote alarm contacts
- Keyed door lock
- Probe access port (⅜")
- HFC-free refrigerant (R600a)
This compact, freestanding undercounter freezer offers a cold-storage solution for limited space in laboratory settings. The unit offers premium features for laboratory storage including a series of temperature alarms to keep stored products at peak effectiveness. Units are equipped with keyed door locks, LED interior lighting, a probe access port, and a microprocessor temperature controller.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty. These freezers run on natural, hydrocarbon refrigerant for environmental cleanliness and economic efficiency.
Material: White cabinet and door
Certifications: UL/C-UL listed
Customers Who Bought This Also Bought