SP Bel-Art Robotic Sterile Pipette Tips, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Designed for use with automated liquid handling equipment, these sterile, disposable, filtered pipette tips can be used in robotic systems as indicated.
- DNase, RNase, pyrogen free; manufactured in Class 8 clean room
- Filtered to prevent contamination of pipette arm
Conductive tip models detect the liquid level when pipetting conductive liquids to ensure repeatable volume accuracy.
Customers Who Bought This Also Bought