Mega Tube Racks for 18 - 20 mm Tubes
Fournisseur: Heathrow Scientific
Certificats
A propos de cet article
High-capacity storage for 15 ml conical and 20 mm round bottom tubes.
- Sturdy and light weight material for easy and secure transport of blood collection tubes
- Alphanumeric identification for quick location and retrieval of tubes
- Place different test groupings into one rack and eliminate need for multiple tube racks
Fully-assembled, polypropylene Mega Tube Racks are great for high throughput labs requiring large sample storage.
Customers Who Bought This Also Bought