J.T.Baker® Plate 96
Fournisseur: Ritter, part of Avantor
Certificats
A propos de cet article
J.T.Baker® Plate 96 round well plates made from PP are ideal for ELISA applications.
- Alphanumeric coding assists with identification of samples
- Plate shape significantly reduces sample carryover
- Standardised dimensions (128×85 mm) ensure compatibility with most common laboratory machines
Spécifications
- Catalog No:
- 43001-0600
- 43001-0118
- 43001-0119
- 43001-0117
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)