J.T.Baker® Plate SRW Magnetic
Supplier: Ritter, part of Avantor
Certificates
About this item
These plates are manufactured from high purity PP for efficient sample preparation and are suitable for use with magnetic particle processors such as the KingFisher™ Flex 96 and Presto.
- Standard SBS footprint
- Cylindrical wells enable good sample mixing
- Slightly raised rims reduce the risk of cross-contamination
- Alphanumeric code assists registration and identification of individual samples
Specifications
- Catalog No:
- 43001-0504
- 43001-0505
- 43001-0506
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)