J.T.Baker® Robotic Tips Type OLA
Fournisseur: Ritter, part of Avantor
Certificats
A propos de cet article
Suitable for use on OLA 2500 and AutoMate 1250/2550 instruments. Available in conductive version, with or without filter.
- 140 tips are sealed in one optimized rack
- Working volumes of 20 - 1200 µl
The validation of the tips on the instrument for specific applications is within the responsibility of the user.
Spécifications
- Catalog No:
- 49044-0100
- 49044-0000
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)