Motic Panthera E2 Upright Compound Microscopes
Fournisseur: Motic
Certificats
A propos de cet article
With the new microscope Series Panthera E2, your decision is distinct. This microscope series allows to experience Life Sciences in a new way, with best possible learning effect.
- Reversed quadruple nosepiece
- LED light indicator
- CCIS Plan UC objectives 4×, 10×, 40× S
- Coaxial coarse and fine focusing adjustment with tension adjustment
- Built in low position coaxial rackless stage with right hand control
- Focusable Abbe condenser N.A.0.90 with iris diaphragm and slot
Panthera E2 microscopes combine high quality materials and precise engineering to a persistent and resilient construction. Smooth operation of the mechanics is self-evident for fast and reliable results, responsible for shifting your daily work in classroom and lab to a higher level of quality. User-friendly design, robust for tough environments and unique features grants the best performance. Models of this series convince positively in smooth operation and haptic.
Ordering information: Contact your Avantor representative to learn more about these and other Motic solutions.
Spécifications
- Catalog No:
- 76480-304
- 76480-310
- 76480-306
- CA76480-306
- 76480-308
- CA76480-310
- CA76480-304
- CA76480-308
- Model:
- Binocular (without 100×)
- Trinocular
- Binocular
- Binocular
- Trinocular (without 100×)
- Trinocular
- Binocular (without 100×)
- Trinocular (without 100×)
- Observation method:
- Darkfield as option, Phase contrast as option, Brightfield
- Darkfield as option, Polarised light, Phase contrast as option, Brightfield
- Darkfield as option, Phase contrast as option, Brightfield
- Darkfield as option, Phase contrast as option, Brightfield
- Darkfield as option, Phase contrast as option, Brightfield
- Darkfield as option, Polarised light, Phase contrast as option, Brightfield
- Darkfield as option, Phase contrast as option, Brightfield
- Darkfield as option, Phase contrast as option, Brightfield
- Head:
- 25° inclined, 360° rotating
- 25° inclined, 360° rotating
- 25° inclined, 360° rotating
- 25° inclined, 360° rotating
- 25° inclined, 360° rotating
- 25° inclined, 360° rotating
- 25° inclined, 360° rotating
- 25° inclined, 360° rotating
- Condenser:
- N.A. 0.90/1.25
- N.A. 0.90/1.25
- N.A. 0.90/1.25
- N.A. 0.90/1.25
- N.A. 0.90/1.25
- N.A. 0.90/1.25
- N.A. 0.90/1.25
- N.A. 0.90/1.25
- Focusing system:
- Coaxial coarse and fine focusing system with tension adjustment
- Coaxial coarse and fine focusing system with tension adjustment
- Coaxial coarse and fine focusing system with tension adjustment
- Coaxial coarse and fine focusing system with tension adjustment
- Coaxial coarse and fine focusing system with tension adjustment
- Coaxial coarse and fine focusing system with tension adjustment
- Coaxial coarse and fine focusing system with tension adjustment
- Coaxial coarse and fine focusing system with tension adjustment
- Illumination:
- Fixed Koehler LED 1W with intensity control
- Fixed Koehler LED 1W with intensity control
- Fixed Koehler LED 1W with intensity control
- Fixed Koehler LED 1W with intensity control
- Fixed Koehler LED 1W with intensity control
- Fixed Koehler LED 1W with intensity control
- Fixed Koehler LED 1W with intensity control
- Fixed Koehler LED 1W with intensity control
- W×D×H:
- 210×282×396 mm
- 210×282×396 mm
- 210×282×396 mm
- 210×282×396 mm
- 210×282×396 mm
- 210×282×396 mm
- 210×282×396 mm
- 210×282×396 mm