ContactUsLinkComponent
Linearity FLQ TDM for Abbott Systems, AUDIT Microcontrols
Linearity FLQ TDM for Abbott Systems, AUDIT Microcontrols
# de catalogue 76485-888
Fournisseur:  Audit Microcontrols
Numéro CAS:  
Linearity FLQ TDM for Abbott System, consisting of five levels of liquid material and additives in human serum, that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments, Storage: 5 days when stored at 2-8CFormat: Frozen Liquid, Size: 5 x 4 mL
Linearity FLQ TDM for Abbott Systems, AUDIT Microcontrols
# de catalogue 76485-888
Fournisseur:  Audit Microcontrols
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Linearity FLQ TDM for Abbott systems
  • Size:
    5 vials
  • Cat. no.:
    76485-888
  • Volume:
    4 ml
Spécifications
A propos de cet article
The Audit® MicroControls™ Linearity FLQ TDM for Abbott systems is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: Acetominophen, Amikacin, Carbamezepine, Digoxin, Gentamicin, Phenobarbital, Phenytoin, Salicylate, Theophylline, Tobramycin, Valproic Acid, Vancomycin.
Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
Customers Who Bought This Also Bought