ABS® Undercounter Pharmacy Refrigerators, Freestanding, Premier Series, Horizon Scientific
Supplier: Horizon Scientific
Certificates
About this item
These Premier Series pharmacy refrigerators are freestanding and space effcient.
- Microprocessor temperature controller
- Digital temperature display
- Audible and visual high and low temperature alarms
- Remote alarm contacts
- Keyed door lock
- Probe access port (³/₈")
- HFC-free refrigerant (R600a)
The Premier Series freestanding refrigerators offer a cold-storage solution to maximize storage in limited space settings. These units offer premium features for pharmacy use including a series of temperature alarms to keep stored products at maximum effectiveness, keyed door locks, shelves for product storage, and temperature controllers to ensure product safety.
Freestanding units require ventilation space on the sides and back. These refrigerators include a two year parts and labor warranty, plus an additional three year compressor parts warranty. They run on HFC-free refrigerant for economic efficiency and environmental safety.
Certifications: UL/C-UL listed.
Specifications
- Catalog No:
- 76478-940
- 76478-942
- 76478-944
- 76478-946
- Model:
- PH-ABT-HC-UCFS-0204
- PH-ABT-HC-UCFS-0204G
- PH-ABT-HC-UCFS-0504
- PH-ABT-HC-UCFS-0504G
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76478-940
- 76478-942
- 76478-944
- 76478-946
- Display:
- Digital
- Digital
- Digital
- Digital
- Defrost:
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Access ports:
- ³/₈" probe
- ³/₈" probe
- ³/₈" probe
- ³/₈" probe
- Shelves/drawers:
- 4 shelves (adjustable)
- 4 shelves (adjustable)
- 3 shelves (adjustable)
- 3 shelves (adjustable)
- Door type:
- 1 solid swing door, right hinged
- 1 glass swing door, right hinged
- 1 solid swing door, right hinged
- 1 glass swing door, right hinged
- Door lock:
- Keyed
- Keyed
- Keyed
- Keyed
- Refrigerant:
- HFC-free refrigerant
- HFC-free refrigerant
- HFC-free refrigerant
- HFC-free refrigerant
- Electrical:
- 115 V, 60 Hz, 0.90 A, 1/8 HP
- 115 V, 60 Hz, 0.90 A, 1/8 HP
- 115 V, 60 Hz, 1.3 A, 1/6 HP
- 115 V, 60 Hz, 1.3 A, 1/6 HP
- Weight:
- 39 kg (85 lbs.)
- 44 kg (97 lbs.)
- 59 kg (129 lbs.)
- 59 kg (129 lbs.)
- Environmental certification:
- Energy Star certified
- Energy Star certified
- Energy Star certified