A propos de cet article
These Premier Series pharmacy freezers are designed to be built into counters for optimal storage space.
- Microprocessor temperature controller
- Digital temperature display
- Audible and visual high and low temperature alarms
- Remote alarm contacts
- Keyed door lock
- Probe access port (³/₈")
- HFC-free refrigerant (R600a)
The Premier Series built in freezers offer a cold-storage solution to maximize storage in limited space settings. These units offer premium features for pharmacy and vaccine use including a series of temperature alarms to keep stored products at maximum effectiveness. Units come equipped with keyed door locks, shelves for product storage, and a temperature monitoring device to ensure product safety. Additionally, units include a pharmacy freezer toolkit and temperature logs for product regulation and control.
These freezers include a two year parts and labor warranty, plus an additional three year compressor parts warranty. They run on HFC-free refrigerant for economic efficiency and environmental safety.
Spécifications
- Catalog No:
- 76478-928
- 76478-926
- Model:
- PH-ABT-HC-UCBI-0420A
- PH-ABT-HC-UCBI-0420
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76478-928
- 76478-926
- Refrigerant:
- HFC-free refrigerant
- HFC-free refrigerant
- Defrost:
- Auto
- Manual
- Electrical:
- 115 V, 60 Hz, 1.8 A, 1/8 HP
- 115 V, 60 Hz, 1.5 A, 1/8 HP
- Access ports:
- ³/₈" probe
- ³/₈" probe