ContactUsLinkComponent
Parter Medical Products Petri Dish, Hardy Diagnostics
Parter Medical Products Petri Dish, Hardy Diagnostics
# de catalogue 76320-910
Fournisseur:  Hardy Diagnostics
Petri Dish, 150x15mm, Manufactured from pure virgin polystyrene
Parter Medical Products Petri Dish, Hardy Diagnostics
# de catalogue 76320-910
Fournisseur:  Hardy Diagnostics
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Petri dish
  • Size:
    150×15 mm
  • Cat. no.:
    76320-910
  • Packaging:
    120 per case
Spécifications
A propos de cet article
The high quality of Parter Medical's Petri dishes is the result of dedication to product excellence, innovative product design and consumer satisfaction.
  • Manufactured from pure virgin polystyrene
  • Quality design and dependable service
Customers Who Bought This Also Bought