VWR® TRIS Glycine-SDS Buffer Powder Packs
Catalog # 76371-728
Supplier: VWR International
CAS Number:
Specifications
- Description:VWR® Tris-Glycine-SDS Buffer
- Size:50 pks
- Cat. no.:76371-728
- pH:8.1 - 8.6
- Formulation:25mM Tris, 192mM Glycine, 0.1% SDS
Specifications
About this item
VWR® Buffer Powder Packs can save time and money, as there is no need to measure and weight many separate chemical components which is required when making buffers from scratch.
- Buffer powder quickly dissolves to make 1L of 1X solution
- No weighing or measuring of separate components
- No pH adjustments required
- Economical shipping, long shelf life
- Quality and consistency guaranteed
VWR® Pre-measured buffer packs are extremely easy to use and are formulated so each pack makes 1 Liter of 1X buffer. Simply tear open a pack and mix completely with 1 liter of distilled water.
Shipping costs are also minimal, as each case of 50 or 100 packs is compact and weighs only 2.5 Lbs.
Certifications: COA avaialble for each lot number
Packaging: Each case of buffer packs is packaged in a convenient dispenser box, which takes up very little lab space. Included in each case is an instruction insert, and a roll of stickers to label each container of prepared buffer solution.
Customers Who Bought This Also Bought