ROX Reference Dye, Biotium
# de catalogue 89138-882
Fournisseur: Biotium
Numéro CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:ROX Passive Reference Dye, 25 uM in TE Buffer
- Size:5 X 1 mL
- Storage temperature:Store at -10 to -35 °C
- Cat. no.:89138-882
- Supplier No.:29052
Spécifications
A propos de cet article
ROX reference dye is used to normalize well-to-well variations that may occur during quantitative, real-time PCR reactions.
- Red dye used to normalize qPCR reactions
- Won’t interfere with PCR reactions
- Supplied as a 25 uM solution
Em/Ex: 600/575 nm
Customers Who Bought This Also Bought