EVERHUTCH NESTCART Mobile Utility Carts, Kewaunee
Fournisseur: Kewaunee
Certificats
A propos de cet article
Mobile units with drawer configurations for storage in clinical and work areas.
- Mobile cart on wheels
- Sitting, ADA, and standing height cabinets
- Drawer configurations
- Under counter storage
- File cabinet
- Supply storage
Ideal for supply storage in clinical areas such as patient rooms. Provides added storage space under a counter to maximize the space. Variable heights allow for sitting, ADA, and standing height work surfaces (30, 33, 36") with full extension drawers. The cart is on four swivel locking casters for easy movement.
Ordering information: The 00 product indicates no locks and 01 indicates all drawers will have a key lock installed.
Customers Who Bought This Also Bought