Eisco LabGlass® Distillation Flasks, 2 Neck with Threaded Joint
Fournisseur: Wards
Certificats
A propos de cet article
High quality distillation flask for chemistry classrooms and laboratories.
- Made of high quality borosilicate glass 3.3
- Ground threaded joints for secure fit with other glassware
- Two neck design
- Autoclavable
- Superior chemical durability
- Low temperature gradient - can withstand high temperatures and thermal shocks
Heat tolerances: strain point 515 °C; annealing point 565 °C and softening point 820 °C.
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Customers Who Bought This Also Bought