Core DX Carts with Hinged Door and Railtop, TrippNT
Fournisseur: TrippNT
Certificats
A propos de cet article
TrippNT's Core DX Cart with Hinged Door and Railtop offer flexible storage with a security railing for additional cart top storage and workspace.
- 1" rail top, 2 locking and 2 swivel 4" twin wheel casters, 3 total locking drawers, 6 side shelves, locking bulk storage with hinged drawer
- Built-in easy to grab handles
- Corrosion-resistant, non-metal HDPE construction
- Organizes, stores, and secures supplies
- 100% total replacement lifetime guarantee/made in USA
TrippNT's Core DX Cart with Hinged Door and Railtop organizes, stores, and secures supplies. It offers a 1" railtop for securesd top storage, 3 drawers, 4 shelves, 2 dispensing shelves, and bulk storage with a hinged clear door. Its built-in handles are ergonomic and easy to grab. It is constructed from a corrosion-resistant non-metal rust proof polymer with robust 4" twin wheel casters capable of easily navigating elevator gaps and uneven surfaces.
Ordering information: Available in 13 color variants.
Delivery: Ships fully assembled.
Customers Who Bought This Also Bought