VWR® Electronic Stem Thermometer, Calibrated, Stainless Steel, 127 mm Probe
Catalog # 98000-156
Supplier: VWR International
Specifications
- Protection class:Waterproof IP 68
- Description:Stem thermometer, field calibratable, maximum temperature indicator, data hold
- Temperature resolution:0.1 °C/°F
- Temperature accuracy:±0.5 °C from 54...+82 °C; ±4 °C above 176 °C; otherwise ±2 °C
- Probe:127 mm (5") stainless steel
- Calibration points:0, 100 °C
- Battery type:LR44
- L×W:178×44 mm (7.0×1.7")
- Cat. no.:98000-156
- Range:–40...+232 ° (–40...+450 ˚F)
Specifications
About this item
Digital stem thermometer is suitable for field studies, wet lab applications, wash down areas, food testing and tanks.
- Will read temperatures in liquids, semi-solids and gases
- Stainless-steel probe easily reaches into cuvettes, test tubes, flasks, beakers and more
- Includes accredited calibration documentation that meets the requirements of ISO 17025
- Individually serialized, CE marked
Certifications: Tested and calibrated in exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST.
Delivery: Includes multi-language instructions (English, French, German, Italian, Portuguese, and Spanish).
Caution: These electronic stem thermometers are not suitable for solids such as frozen items.
Customers Who Bought This Also Bought