Epredia™ Premium Microscope Slides in Tropical Packaging, Erie Scientific (Thermo Scientific)
Fournisseur: Erie Scientific
Certificats
A propos de cet article
Microscope slides are packaged in a sealed tropical barrier bag with desiccant.
- Manufactured from pure white glass
- Reduced iron content for low autofluorescence
- Precleaned for immediate use
- Non-adhesion style slides
Epredia™ premium microscope slides are supplied in a sealed tropical barrier bag with desiccant to reduce slide surface deterioration and sticking. Manufactured in the USA.
Customers Who Bought This Also Bought