Motic Moticam S Series, Microscope Cameras
Fournisseur: National Optical
Certificats
A propos de cet article
Ideal for in-person or distance learning and instruction.
- Fast frame rates
- sCMOS sensor
- USB 3.1 interface
Motic's new C-mount ready sCMOS cameras are the newest members of the Moticam family of professionally designed and engineered cameras. Easy enough for the microscopy novice, but sophisticated enough for the professional. With back illuminated sensor technology and speedy data transfer, this camera creates superbly detailed images and smooth video captures.
Ordering information: Included Motic Images software enables live and captured images at a maximum quality. Edit, measure, and share images or reports with this comprehensive software.
Customers Who Bought This Also Bought