ContactUsLinkComponent
Rapid Response® Plastic Collection Cup, BTNX
Rapid Response® Plastic Collection Cup, BTNX
# de catalogue 76473-384
Fournisseur:  BTNX INC.
Collection Cup, Plastic(Polypropylene), for clarity and durability and is ideal for sample collection.
Rapid Response® Plastic Collection Cup, BTNX
# de catalogue 76473-384
Fournisseur:  BTNX INC.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Capacity:
    90 ml
  • Description:
    Plastic collection cup
  • Cat. no.:
    76473-384
Spécifications
A propos de cet article
The Rapid Response® Plastic collection cup is a non sterile cup made with polypropylene allowing for clarity and durability and is ideal for sample collection.
  • 90 ml volume
  • 45 mm diameter
  • Screw-on cap
The cup includes graduations on the cup exterior for easy reading and made with polypropylene material for clarity and durability when reviewing samples. The compact size is ideal for sample testing with strips, cassettes, and 1 - 12 panel drug test.
Delivery: Includes collection cups.
Customers Who Bought This Also Bought