A propos de cet article
The Rapid Response® Multi-drug saliva test cup is a rapid, qualitative screening test for the simultaneous detection of multiple drugs and metabolites in human oral fluid. An indicator strip is included in the indicator window of the collection swab to signal saturation. Upon saturation, the collection swab will change colour.
- Sample: Saliva
- Format: Cup
- Time to result: 10 minutes
- Storage Condition: 2 to 30 °C (36 to 86 °F)
- Test principle: Lateral flow immunoassay
Interpretation of test results are: positive (one line), negative (two lines), invalid (no lines or no Control line); or the comparison of reaction pad color to the corresponding color blocks for the adulteration strips.
Multi-Drug Urine Test Cups can include THC (40), COC (20), OPI (40), AMP (50), MET (50), BAR (50), BZO (10), MDMA (50), MTD (30), PCP (10), OXY (20), FYL (10), K2 (50), BUP (5), OPI (25), PCP (10), AMP (1000), BUP (10), BZO (300), COC (300), MTD (300), MET (1000), OPI (2000), OXY (100), THC (50).