A propos de cet article
The Rapid Response® Multi-drug test panel is a rapid, one-step, qualitative test for the simultaneous detection of multiple drugs and metabolites in human urine.
- Sample: Urine
- Format: Panel
- Time to result: 5 minutes
- Storage Condition: 2 to 30 °C (36 to 86 °F)
- Test principle: Lateral flow immunoassay
Multi-Drug Test Panels can include COC (300), AMP (1000), MET (1000), THC (50), MTD (300), OPI (2000), PCP (25), BAR (300), BZO (300), TCA (1000), MDMA (500), MOP (300), OXY (100), PPX (300), BUP (10), FYL (10).