ContactUsLinkComponent
Oakton® Printer Paper, RS232 Microprinters for 2700 Meters, Cole Parmer
Oakton® Printer Paper, RS232 Microprinters for 2700 Meters, Cole Parmer
Catalog # 76221-230
Supplier:  Argos Technologies
Printer Paper 2 Rolls 2.28x1.81x72 inch
Oakton® Printer Paper, RS232 Microprinters for 2700 Meters, Cole Parmer
Catalog # 76221-230
Supplier:  Argos Technologies
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • For Use With:
    Oakton® thermal printer with RS232 for 2700 meters
  • Description:
    Oakton® printer paper, 2 rolls
  • Cat. no.:
    76221-230
Specifications
About this item
Convenient printer for Oakton 2700 meters.
  • Dot-matrix printers
  • Real-time clock available
Offers the convenience of directly printing from your meter. This compact dot-matrix microprinter connects easily to your benchtop meter via an RS232C outlet - ideal for data -recording and report-generation. Uses paper-rolls (2-rolls paper).
Ordering information: Printer paper only; printer sold separately.
Customers Who Bought This Also Bought